Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SESTD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585032
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585032 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585032 Supplier Invitrogen™ Supplier No. PA585032
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WNVVKTVVVM LQNVVPAEVS LVCVVKPDEF WDKKVTHFCF WKEKDRLGFE VILVSANKLT RYIEPCQLTE DFGGSL Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 99%.

SESTD1 was initially identified in mutant zebrafish with defects in the spontaneous contraction and touch response as a novel gene, solo, encoding a protein containing SEC14 and spectrin repeat domains. Other experiments indicated that SESTD1 interacts with the TRPC4 and TRPC5, members of the transient receptor potential channel family, via the TRPC calmodulin- and inositol 1, 4, 5-triphosphate receptor-binding domain and is essential for efficient receptor-mediated activation of TRPC5, suggesting that SESTD1 is a novel regulator of these TRPC proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SESTD1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Sestd1
Gene Accession No. Q86VW0
Gene Alias 1500031J16Rik; A930010O20; EGK_04585; Huntingtin-interacting protein-like protein; protein Solo; RGD1562244; SEC14 and spectrin domain containing 1; SEC14 and spectrin domains 1; SEC14 domain and spectrin repeat-containing protein 1; Sestd1; SOLO; Solo/Trio8
Gene Symbols Sestd1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SESTD1. Recombinant protein control fragment (Product #RP-108680).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91404
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.