Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SET Antibody (PCRP-SET-1C6), PerCP, Novus Biologicals™
SDP

Catalog No. NB378908 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB378908 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB378908 Supplier Novus Biologicals Supplier No. NBP308730PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SET Monoclonal antibody specifically detects SET in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array

Specifications

Antigen SET
Applications Western Blot, Flow Cytometry, Immunofluorescence, Protein Array
Classification Monoclonal
Clone PCRP-SET-1C6
Conjugate PerCP
Formulation PBS
Gene Alias 2PP2A, HLA-DR-associated protein II, I2PP2A, I-2PP2A, IGAAD, Inhibitor of granzyme A-activated DNase, inhibitor-2 of protein phosphatase-2A, IPP2A2, PHAPIITAF-I, Phosphatase 2A inhibitor I2PP2A, protein phosphatase type 2A inhibitor, protein SET, SET nuclear oncogene, SET translocation (myeloid leukemia-associated), TAF-IBETA, Template-activating factor I, Template-Activating Factor-I, chromatin remodelling factor
Host Species Mouse
Immunogen Recombinant full-length human SET protein (Uniprot: Q01105 )
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6418
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.