Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SETD1A Antibody (OTI7B7), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP274144A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP274144A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP274144A32 Supplier Novus Biologicals Supplier No. NBP274144AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SETD1A Monoclonal specifically detects SETD1A in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen SETD1A
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI7B7
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias histone-lysine N-methyltransferase SETD1A, hSET1A, KIAA0339EC 2.1.1.43, KMT2FSET1, Lysine N-methyltransferase 2F, SET domain containing 1A, SET domain-containing protein 1A, Set1, Set1/Ash2 histone methyltransferase complex subunit SET1, SET1A
Gene Symbols SETD1A
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-320 of human SETD1A (NP_055527) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 9739
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.