Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SETD3 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB289968 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289968 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289968 Supplier Novus Biologicals Supplier No. NBP232136PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SETD3 Polyclonal antibody specifically detects SETD3 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen SETD3
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias C14orf154, chromosome 14 open reading frame 154, DKFZp761E1415, EC 2.1.1.43, FLJ23027, MGC87236, SET domain containing 3, SET domain-containing protein 3
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human SET Domain Containing 3 using the numbering given in entry NP_115609.2 (GeneID 84193).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84193
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.