Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SETD7/9 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

$206.50 - $426.00

Specifications

Antigen SETD7/9
Dilution Western Blot 1:500-1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:100, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ChIP Assay
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056664T
SDP
View Documents
Novus Biologicals
NB10056664SS
0.05 mL
Each for $206.50
Only null left
Add to Cart
 
NB10056664
SDP
View Documents
Novus Biologicals
NB10056664
0.2 mL
Each for $426.00
Only null left
Add to Cart
 
Description

Description

SETD7/9 Polyclonal specifically detects SETD7/9 in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

SETD7/9
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ChIP Assay
Unconjugated
Rabbit
Histones and Modified Histones, Signal Transduction
Neat whole antisera with 0.05% Sodium Azide
EC 2.1.1.43, H3-K4-HMTase SETD7, histone H3-lysine 4-specific methyltransferase, histone-lysine N-methyltransferase SETD7, KIAA1717SET domain-containing protein 7, KMT7FLJ21193, Lysine N-methyltransferase 7, SET domain containing (lysine methyltransferase) 7, SET7/9Histone H3-K4 methyltransferase SETD7, SET7Set9, SET9
SETD7
IgG
Unpurified
This is specific for Set7/9 (a histone H3 lysine 4 methyltransferase) and does not recognize PR-Set7 (a histone H4 lysine 20 methyltransferase).
Western Blot 1:500-1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:100, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Polyclonal
Antisera
RUO
Human, Mouse
Q8WTS6
80854
Two synthethic peptides corresponding to amino acids 131-145 (GEVNEDGEMTGEKIA) and 336-352 (GYDHSPPGKSGPEAPEW) of human SETD7 were used as immunogen for this antibody.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.