Learn More
Description
Specifications
Specifications
| Antigen | SETD7/9 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Dilution | Western Blot, ELISA |
| Formulation | PBS |
| Gene Alias | EC 2.1.1.43, H3-K4-HMTase SETD7, histone H3-lysine 4-specific methyltransferase, histone-lysine N-methyltransferase SETD7, KIAA1717SET domain-containing protein 7, KMT7FLJ21193, Lysine N-methyltransferase 7, SET domain containing (lysine methyltransferase) 7, SET7/9Histone H3-K4 methyltransferase SETD7, SET7Set9, SET9 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant SETD7/9 (NP_085151.1; Asp2-Lys366). |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
