Learn More
Description
Specifications
Specifications
| Antigen | SETD8 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | DyLight 488 |
| Dilution | Western Blot 1:1000, Immunohistochemistry (Negative), ICC/IF (Negative) |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Accession No. | Q9NQR1 |
| Gene Alias | H4-K20-HMTase SETD8, H4K20-specific histone methyltransferase splice variant Set8b, Histone-lysine N-methyltransferase SETD8, KMT5A, KMT5A SET domain-containing protein 8, Lysine N-methyltransferase 5A, PR/SET domain containing protein 8, PR/SET domain-containing protein 07, PR/SET07, PRSET7, PR-Set7EC 2.1.1.43, SET domain containing (lysine methyltransferase) 8, SET07SET8H4-K20-specific histone methyltransferase, SET8 |
| Gene Symbols | SETD8 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
