Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SETD8 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB063247 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB063247 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB063247 Supplier Novus Biologicals Supplier No. NBP299864H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SETD8 Polyclonal specifically detects SETD8 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen SETD8
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate HRP
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias H4-K20-HMTase SETD8, H4K20-specific histone methyltransferase splice variant Set8b, Histone-lysine N-methyltransferase SETD8, KMT5A, KMT5A SET domain-containing protein 8, Lysine N-methyltransferase 5A, PR/SET domain containing protein 8, PR/SET domain-containing protein 07, PR/SET07, PRSET7, PR-Set7EC 2.1.1.43, SET domain containing (lysine methyltransferase) 8, SET07SET8H4-K20-specific histone methyltransferase, SET8
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant SETD8 (NP_065115.3; Lys195-His352).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 387893
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.