Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SETDB2 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001137 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001137 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001137 Supplier Novus Biologicals Supplier No. NB1001137
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 2 publications

SETDB2 Polyclonal specifically detects SETDB2 in Human samples. It is validated for Chemotaxis, ChIP Assay, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Peptide ELISA, Western Blot.

Specifications

Antigen SETDB2
Applications Chemotaxis, ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/mL, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Peptide ELISA Detection limit 1:1000, Chromatin Immunoprecipitation (ChIP)
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_114121.
Gene Alias C13orf4, chromosome 13 open reading frame 4, chronic lymphocytic leukemia deletion region 8, Chronic lymphocytic leukemia deletion region gene 8 protein, CLLD8DKFZp761J1217, DKFZp586I0123, EC 2.1.1.43, histone-lysine N-methyltransferase SETDB2, KMT1FCLLL8, Lysine N-methyltransferase 1F, SET domain bifurcated 2, SET domain, bifurcated 2
Gene Symbols SETDB2
Host Species Goat
Immunogen Peptide with sequence GEKNGDAKTFWME-C corresponding to N-Terminus according to NP_114121.2, NP_001153780.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83852
Test Specificity This antibody is expected to recognize both reported isoforms (NP_114121.2; NP_001153780.1).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.