Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SETDB2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP131264 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP131264 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP131264 Supplier Novus Biologicals Supplier No. NBP131264
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SETDB2 Polyclonal specifically detects SETDB2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SETDB2
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1:100-1:2000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Gene Accession No. Q96T68
Gene Alias C13orf4, chromosome 13 open reading frame 4, chronic lymphocytic leukemia deletion region 8, Chronic lymphocytic leukemia deletion region gene 8 protein, CLLD8DKFZp761J1217, DKFZp586I0123, EC 2.1.1.43, histone-lysine N-methyltransferase SETDB2, KMT1FCLLL8, Lysine N-methyltransferase 1F, SET domain bifurcated 2, SET domain, bifurcated 2
Gene Symbols SETDB2
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human SETDB2. The exact sequence is proprietary.
Molecular Weight of Antigen 82 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83852
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.