Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SF-1/NR5A1/Steroidogenic Factor 1 Antibody (NR5A1/3397), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB385490 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB385490 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB385490 Supplier Novus Biologicals Supplier No. NBP308624IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SF-1/NR5A1/Steroidogenic Factor 1 Monoclonal antibody specifically detects SF-1/NR5A1/Steroidogenic Factor 1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen SF-1/NR5A1/Steroidogenic Factor 1
Applications Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone NR5A1/3397
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias AD4BPSTF-1, Adrenal 4-binding protein, ELP, FTZ1adrenal 4 binding protein, FTZF1nuclear receptor AdBP4, Fushi tarazu factor homolog 1, Nuclear receptor subfamily 5 group A member 1, nuclear receptor subfamily 5, group A, member 1, SF-1POF7, SF1steroidogenic factor 1, Steroid hormone receptor Ad4BP, steroidogenic factor-1
Host Species Mouse
Immunogen Recombinant fragment (around aa220-461) of human SF-1/NR5A1/Steroidogenic Factor 1 protein (Uniprot: Q13285)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 2516
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.