Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SF20/MYDGF Antibody (2562A), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB339869 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339869 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339869 Supplier Novus Biologicals Supplier No. FAB1104MFV500
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SF20/MYDGF Monoclonal specifically detects SF20/MYDGF in Human, Mouse, Rat samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunohistochemistry, Western Blot.

Specifications

Antigen SF20/MYDGF
Applications CyTOF, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2562A
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias C19orf10, chromosome 19 open reading frame 10, EUROIMAGE1875335, IL25, IL27, IL27w, interleukin 27 working designation, interleukin-25, Ly6elg, MYDGF, myeloid-derived growth factor, R33729_1, SF20, Stromal cell-derived growth factor SF20, UPF0556 protein C19orf10
Host Species Rabbit
Immunogen Mouse myeloma cell line NS0-derived mouse SF20/MYDGF, Val25-Leu166, Accession # Q9CPT4
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 56005
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.