Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SF3A1 Antibody (OTI3F4), Biotin, Novus Biologicals™
SDP

Catalog No. NBP274150B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274150B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274150B Supplier Novus Biologicals Supplier No. NBP274150B

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

SF3A1 Monoclonal specifically detects SF3A1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen SF3A1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3F4
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias pre-mRNA processing 21, pre-mRNA splicing factor SF3a (120 kDa subunit), Prp21, PRPF21, SAP 114, SAP114splicing factor 3a, subunit 1, 120kD, SF3a120, spliceosome associated protein 114, Spliceosome-associated protein 114, splicing factor 3 subunit 1, splicing factor 3A subunit 1, splicing factor 3a, subunit 1, 120kDa
Gene Symbols SF3A1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 249-568 of human SF3A1 (NP_005868) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 10291.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.