Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SF3B14 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15722720 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15722720 20 μL
NBP157227 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15722720 Supplier Novus Biologicals Supplier No. NBP15722720UL

Rabbit Polyclonal Antibody

SF3B14 Polyclonal specifically detects SF3B14 in Human samples. It is validated for Western Blot.

Specifications

Antigen SF3B14
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS & 2% Sucrose. with No Preservative
Gene Accession No. Q7RTV0
Gene Alias HSPC175, Ht006, P14, pre-mRNA branch site protein p14, SAP14, SF3b 14 kDa subunit, SF3B14a, spliceosome-associated protein, 14 kDa subunit, splicing factor 3B, 14 kDa subunit
Gene Symbols SF3B14
Host Species Rabbit
Immunogen Synthetic peptides corresponding to SF3B14(splicing factor 3B, 14 kDa subunit) The peptide sequence was selected from the middle region of SF3B14. Peptide sequence HLSGFNVCNRYLVVLYYNANRAFQKMDTKKKEEQLKLLKEKYGINTDPPK.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51639
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.