Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SF3B2 Rabbit anti-Human, Alexa Fluor 594, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB102995 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB102995 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB102995 Supplier Novus Biologicals Supplier No. NBP297657AF594

Rabbit Polyclonal Antibody

SF3B2 Polyclonal specifically detects SF3B2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SF3B2
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50 mM Sodium Borate
Gene Alias Cus1, pre-mRNA splicing factor SF3b 145 kDa subunit, Pre-mRNA-splicing factor SF3b 145 kDa subunit, SAP145SAP 145, SF3b1, SF3b145, SF3b150, spliceosome associated protein 145, Spliceosome-associated protein 145, splicing factor 3B subunit 2, splicing factor 3b, subunit 2, 145kD, splicing factor 3b, subunit 2, 145kDa
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SF3B2 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10992
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.