Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SF3B4 Monoclonal Antibody (3A1)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA547407

 View more versions of this product

Catalog No. PIMA547407


Only null left
Add to Cart

Description

Description

SF3B4 Monoclonal Antibody for Western Blot, ICC/IF, IHC

This gene encodes one of four subunits of the splicing factor 3B. The protein encoded by this gene cross-links to a region in the pre-mRNA immediately upstream of the branchpoint sequence in pre-mRNA in the prespliceosomal complex A. It also may be involved in the assembly of the B, C and E spliceosomal complexes. In addition to RNA-binding activity, this protein interacts directly and highly specifically with subunit 2 of the splicing factor 3B. This protein contains two N-terminal RNA-recognition motifs (RRMs), consistent with the observation that it binds directly to pre-mRNA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SF3B4
Monoclonal
1 mg/mL
PBS with 50% glycerol and 5mM sodium azide
Q15427, Q6AYL5, Q8QZY9
Sf3b4
Full length recombinant human SF3B4 expressed in and purified from E. coli.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG2b
Immunohistochemistry, Western Blot, Immunocytochemistry
3A1
Unconjugated
Sf3b4
49kD; AFD1; chunp6902; fb36a04; Hsh49; MGC10828; pre-mRNA-splicing factor SF3b 49 kDa subunit; SAP 49; Sap49; SAP49MGC10828; sf3b4; SF3b49; SF3b50; spliceosomal protein; spliceosome-associated protein (U2 snRNP); spliceosome-associated protein 49; Splicing factor 3B subunit 4; splicing factor 3b, subunit 4; splicing factor 3b, subunit 4, 49kD; splicing factor 3b, subunit 4, 49kDa; Unknown (protein for IMAGE:8120437); wu:fb36a04
Mouse
Protein A/G
RUO
10262, 107701, 295270
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.