Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SF3B4 Monoclonal Antibody (3A1)
Mouse Monoclonal Antibody
$526.00 - $1800.00
Specifications
Antigen | SF3B4 |
---|---|
Clone | 3A1 |
Concentration | 1 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA547309
|
Invitrogen™
MA547309 |
500 μL |
Each for $1,800.00
|
|
|||||
PIMA547407
|
Invitrogen™
MA547407 |
100 μL |
Each for $526.00
|
|
|||||
Description
SF3B4 Monoclonal Antibody for Western Blot, ICC/IF, IHC
This gene encodes one of four subunits of the splicing factor 3B. The protein encoded by this gene cross-links to a region in the pre-mRNA immediately upstream of the branchpoint sequence in pre-mRNA in the prespliceosomal complex A. It also may be involved in the assembly of the B, C and E spliceosomal complexes. In addition to RNA-binding activity, this protein interacts directly and highly specifically with subunit 2 of the splicing factor 3B. This protein contains two N-terminal RNA-recognition motifs (RRMs), consistent with the observation that it binds directly to pre-mRNA.Specifications
SF3B4 | |
1 mg/mL | |
Immunohistochemistry, Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 5mM sodium azide | |
Q15427, Q6AYL5, Q8QZY9 | |
10262, 107701, 295270 | |
Full length recombinant human SF3B4 expressed in and purified from E. coli. | |
Antibody |
3A1 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Monoclonal | |
Liquid | |
IgG2b | |
Human, Mouse, Rat | |
Sf3b4 | |
49kD; AFD1; chunp6902; fb36a04; Hsh49; MGC10828; pre-mRNA-splicing factor SF3b 49 kDa subunit; SAP 49; Sap49; SAP49MGC10828; sf3b4; SF3b49; SF3b50; spliceosomal protein; spliceosome-associated protein (U2 snRNP); spliceosome-associated protein 49; Splicing factor 3B subunit 4; splicing factor 3b, subunit 4; splicing factor 3b, subunit 4, 49kD; splicing factor 3b, subunit 4, 49kDa; Unknown (protein for IMAGE:8120437); wu:fb36a04 | |
Sf3b4 | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title