Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SF3B4 Rabbit anti-Human, DyLight 350, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB046212 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB046212 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB046212 Supplier Novus Biologicals Supplier No. NBP297663UV

Rabbit Polyclonal Antibody

SF3B4 Polyclonal specifically detects SF3B4 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SF3B4
Applications Immunofluorescence
Classification Polyclonal
Conjugate DyLight 350
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Hsh49, pre-mRNA splicing factor SF3b 49 kDa subunit, Pre-mRNA-splicing factor SF3b 49 kDa subunit, SAP 49, SAP49MGC10828, SF3B49, SF3b50, spliceosomal protein, spliceosome-associated protein (U2 snRNP), Spliceosome-associated protein 49, splicing factor 3B subunit 4, splicing factor 3b, subunit 4, 49kD, splicing factor 3b, subunit 4, 49kDa
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SF3B4 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 10262
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.