Learn More
SFMBT1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | SFMBT1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-4576
![]() |
Bethyl Laboratories, Inc
A303221AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Rat
SFMBT1 (Scm-like with four MBT domains 1) gene shares high similarity with the Drosophila Scm (sex comb on midleg) gene. It encodes a protein which contains four maligt brain tumor repeat (MBT) domains, and may be involved in antigen recognition. Several alternative splice variants that encode the same protein have been characterized.Specifications
SFMBT1 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
RU1, SFMBT, Scm-related gene containing four mbt domains, Scm-related gene product containing four mbt domains, renal ubiquitous protein 1, scm-like with four MBT domains protein 1 | |
SFMBT1 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9UHJ3 | |
51460 | |
Between 816 and 866 | |
Primary | |
4° C | |
SFMBT1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.