Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SFPQ Antibody (B92) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB12011825 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB12011825 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB12011825 Supplier Novus Biologicals Supplier No. NB12011825
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SFPQ Monoclonal specifically detects SFPQ in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen SFPQ
Applications Western Blot
Classification Monoclonal
Clone B92
Concentration 2.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-4 ug/ml
Gene Alias 100 kDa DNA-pairing protein, DNA-binding p52/p100 complex, 100 kDa subunit, hPOMp100, polypyrimidine tract binding protein associated, polypyrimidine tract-binding protein-associated splicing factor, Polypyrimidine tract-binding protein-associated-splicing factor, PSFPOMP100, PTB-associated splicing factor, PTB-associated-splicing factor, splicing factor proline/glutamine rich (polypyrimidine tract binding proteinassociated), splicing factor proline/glutamine rich (polypyrimidine tract-bindingprotein-associated), splicing factor proline/glutamine-rich, splicing factor, proline- and glutamine-rich
Gene Symbols SFPQ
Host Species Mouse
Immunogen Tissue cell preparation (lysate of mouse bone marrow-derived stromal cell line.).
Purification Method Protein A or G purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline DNA Repair, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 6421
Test Specificity SFPQ (B92)
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.