Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sFRP-1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB325604 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB325604 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB325604 Supplier Novus Biologicals Supplier No. NBP299417MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

sFRP-1 Polyclonal antibody specifically detects sFRP-1 in Mouse samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen sFRP-1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias FRP1, FRP-1FrzA, FRPSARP-2, SARP2sFRP-1, Secreted apoptosis-related protein 2, secreted frizzled-related protein 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human sFRP-1
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 6422
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.