Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sFRP-1 Antibody, Novus Biologicals™
SDP

Catalog No. NB600499 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600499 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600499 Supplier Novus Biologicals Supplier No. NB600499
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

sFRP-1 Polyclonal specifically detects sFRP-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen sFRP-1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:200-1:2000, ELISA 1:5000-1:15000, Immunohistochemistry 1:800, Immunocytochemistry/Immunofluorescence 5 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. Q8N474
Gene Alias FRP1, FRP-1FrzA, FRPSARP-2, SARP2sFRP-1, Secreted apoptosis-related protein 2, secreted frizzled-related protein 1
Gene Symbols SFRP1
Host Species Rabbit
Immunogen sFRP-1 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a 12 aa region of human Sfrp1 protein. (Uniprot: Q8N474)
Molecular Weight of Antigen 35.39 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 6422
Test Specificity SFRP1
Target Species Human
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.