Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sFRP-2 Antibody, Novus Biologicals™
SDP

Catalog No. NB171868 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171868 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB171868 Supplier Novus Biologicals Supplier No. NBP318244
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

sFRP-2 Polyclonal antibody specifically detects sFRP-2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen sFRP-2
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:50
Formulation PBS pH 7.4, 50% glycerol
Gene Alias FRP2, FRP-2secreted apoptosis related protein 1, SARP-1, SARP1sFRP-2, SDF-5, Secreted apoptosis-related protein 1, secreted frizzled-related protein 2
Host Species Rabbit
Immunogen Synthetic peptide from the C-terminal of Human Secreted frizzled-related protein 2 (aa. 195-295)
Purification Method Peptide affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6423
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.