Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sFRP-4 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB081383 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB081383 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB081383 Supplier Novus Biologicals Supplier No. NBP298217B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

sFRP-4 Polyclonal specifically detects sFRP-4 in Mouse samples. It is validated for ELISA.

Specifications

Antigen sFRP-4
Applications ELISA
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias Frizzled protein, human endometrium, FRP-4, frpHE, FRPHEMGC26498, secreted frizzled-related protein 4, secreted frizzled-related protein 4; secreted frizzled-related protein 4, sFRP-4
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant sFRP-4 (NP_057896.1; Met 1-Ser 351).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Growth and Development, Neuronal Cell Markers, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 6424
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.