Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sFRP-4 Rabbit anti-Human, Mouse, Rat, Clone: S01-2C2, Novus Biologicals™
SDP

Catalog No. NB188305 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB188305 25 μg
NB188304 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB188305 Supplier Novus Biologicals Supplier No. NBP31497425UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

sFRP-4 Monoclonal antibody specifically detects sFRP-4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin), Immunofluorescence

Specifications

Antigen sFRP-4
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone S01-2C2
Conjugate Unconjugated
Dilution Western Blot, ELISA, Immunohistochemistry-Paraffin 20 μg/mL, Immunofluorescence 20 μg/mL
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide
Gene Alias Frizzled protein, human endometrium, FRP-4, frpHE, FRPHEMGC26498, secreted frizzled-related protein 4, secreted frizzled-related protein 4; secreted frizzled-related protein 4, sFRP-4
Host Species Rabbit
Immunogen A synthetic peptide of human sFRP-4 (Uniprot#: Q6FHJ7)
Purification Method Affinity-purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Growth and Development, Neuronal Cell Markers, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 6424
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.