Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SFRS12IP1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP154710 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP154710 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP154710 Supplier Novus Biologicals Supplier No. NBP154710
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SFRS12IP1 Polyclonal specifically detects SFRS12IP1 in Human samples. It is validated for Western Blot.

Specifications

Antigen SFRS12IP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q8N9Q2
Gene Alias MGC131910, p18 splicing regulatory protein, p18SRP, P18SRPMGC150549, protein SFRS12IP1, protein SREK1IP1, SFRS12-interacting protein 1FLJ36754, SFRS12IP1, Splicing regulatory protein of 18 kDa, SREK1-interacting protein 1MGC150548
Gene Symbols SREK1IP1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to SFRS12IP1(SFRS12-interacting protein 1) The peptide sequence was selected from the middle region of SFRS12IP1. Peptide sequence NEEEEKKKEKSKEKIKLKKKRKRSYSSSSTEEDTSKQKKQKYQKKEKKKE.
Molecular Weight of Antigen 18 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285672
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.