Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SFRS12IP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | SFRS12IP1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
SFRS12IP1 Polyclonal specifically detects SFRS12IP1 in Human samples. It is validated for Western Blot.Specifications
SFRS12IP1 | |
Polyclonal | |
Rabbit | |
Q8N9Q2 | |
285672 | |
Synthetic peptides corresponding to SFRS12IP1(SFRS12-interacting protein 1) The peptide sequence was selected from the middle region of SFRS12IP1. Peptide sequence NEEEEKKKEKSKEKIKLKKKRKRSYSSSSTEEDTSKQKKQKYQKKEKKKE. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
MGC131910, p18 splicing regulatory protein, p18SRP, P18SRPMGC150549, protein SFRS12IP1, protein SREK1IP1, SFRS12-interacting protein 1FLJ36754, SFRS12IP1, Splicing regulatory protein of 18 kDa, SREK1-interacting protein 1MGC150548 | |
SREK1IP1 | |
IgG | |
18 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title