Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SFRS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | SFRS2 |
---|---|
Concentration | 0.3 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-557-097
![]() ![]() |
Proteintech Group Inc
203711AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The antibody has no cross reaction to SFRS2B.
SFRS2 is necessary for the splicing of pre-mRNA. The protein is required for formation of the earliest ATP-dependent splicing complex and interacts with spliceosomal components bound to both the 5′- and 3′-splice sites during spliceosome assembly. It also is required for ATP-dependent interactions of both U1 and U2 snRNPs with pre-mRNA. And it interacts with other spliceosomal components, via the RS domains, to form a bridge between the 5′- and 3′-splice site binding components, U1 snRNP and U2AF. It binds to purine-rich RNA sequences, either 5′-AGSAGAGTA-3′ (S=C or G) or 5′-GTTCGAGTA-3′ and can bind to beta-globin mRNA and commit it to the splicing pathway.Specifications
SFRS2 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q01130, Q62093, Q6PDU1 | |
20382, 494445, 6427 | |
Peptide | |
Primary | |
-20°C | |
SRSF2 |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
PR264, Protein PR264, SC 35, SC35, SFRS2, SFRS2A, Splicing component, 35 kDa, Splicing factor SC35, SRp30b, SRSF2 | |
Srsf2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title