Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SFRS2B Antibody, Novus Biologicals™
SDP

Catalog No. NBP15716820 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15716820 20 μL
NBP157168 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15716820 Supplier Novus Biologicals Supplier No. NBP15716820UL

Rabbit Polyclonal Antibody

SFRS2B Polyclonal specifically detects SFRS2B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SFRS2B
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry
Formulation PBS & 2% Sucrose. with No Preservative
Gene Alias Pre-mRNA-splicing factor SRP46, serine/arginine-rich splicing factor 8, Splicing factor SRp46, Splicing factor, arginine/serine-rich 2Bsplicing factor, arginine/serine-rich, 46kD, SR splicing factor 8, SRP46SFRS2B
Gene Symbols SRSF8
Host Species Rabbit
Immunogen Synthetic peptides corresponding to SFRS2B(splicing factor, arginine/serine-rich 2B) The peptide sequence was selected from the middle region of SFRS2B. Peptide sequence YRESRYGGSHYSSSGYSNSRYSRYHSSRSHSKSGSSTSSRSASTSKSSSA.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10929
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.