Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SFRS2B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$158.00 - $487.50
Specifications
Antigen | SFRS2B |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Regulatory Status | RUO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15716820
![]() |
Novus Biologicals
NBP15716820UL |
20 μL |
Each for $158.00
|
|
|||||
NBP157168
![]() |
Novus Biologicals
NBP157168 |
100 μL |
Each for $487.50
|
|
|||||
Description
SFRS2B Polyclonal specifically detects SFRS2B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SFRS2B | |
Unconjugated | |
RUO | |
Pre-mRNA-splicing factor SRP46, serine/arginine-rich splicing factor 8, Splicing factor SRp46, Splicing factor, arginine/serine-rich 2Bsplicing factor, arginine/serine-rich, 46kD, SR splicing factor 8, SRP46SFRS2B | |
SRSF8 | |
IgG |
Polyclonal | |
Rabbit | |
Human | |
10929 | |
Synthetic peptides corresponding to SFRS2B(splicing factor, arginine/serine-rich 2B) The peptide sequence was selected from the middle region of SFRS2B. Peptide sequence YRESRYGGSHYSSSGYSNSRYSRYHSSRSHSKSGSSTSSRSASTSKSSSA. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title