Learn More
SFRS3 Antibody - BSA Free, Novus Biologicals™
SDP
Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals NBP241316
 View more versions of this product
Catalog No. NBP241316
Only null left
Add to Cart
Description
Description
SFRS3 Polyclonal specifically detects SFRS3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
SFRS3
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
SRSF3
Antibody was raised against a 19 amino acid peptide near the center of human SRSF3. The immunogen is located within amino acids 80 - 130 of SRSF3. Amino Acid Squence: PSWGRRPRDDYRRRSPPPR
Affinity Purified
RUO
6428
Human, Mouse, Rat
IgG
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
1 mg/mL
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunocytochemistry
Pre-mRNA-splicing factor SRP20, serine/arginine-rich splicing factor 3, SFRS3splicing factor, arginine/serine-rich, 20-kD, Splicing factor, arginine/serine-rich 3SRP20, SRp20
Rabbit
19 kDa
0.1 mg
Primary
SRSF3 antibody is human, mouse and rat reactive.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.