Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SFRS3 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $499.50
Specifications
Antigen | SFRS3 |
---|---|
Concentration | 1 mg/mL |
Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SFRS3 Polyclonal specifically detects SFRS3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
SFRS3 | |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
6428 | |
Antibody was raised against a 19 amino acid peptide near the center of human SRSF3. The immunogen is located within amino acids 80 - 130 of SRSF3. Amino Acid Squence: PSWGRRPRDDYRRRSPPPR | |
Primary | |
SRSF3 antibody is human, mouse and rat reactive. |
1 mg/mL | |
Polyclonal | |
Rabbit | |
Human, Mouse, Rat | |
Pre-mRNA-splicing factor SRP20, serine/arginine-rich splicing factor 3, SFRS3splicing factor, arginine/serine-rich, 20-kD, Splicing factor, arginine/serine-rich 3SRP20, SRp20 | |
SRSF3 | |
IgG | |
Affinity Purified | |
19 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title