Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SFTS Virus HB29 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB343929 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343929 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343929 Supplier Novus Biologicals Supplier No. NBP241155MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SFTS Virus HB29 Polyclonal antibody specifically detects SFTS Virus HB29 in Virus samples. It is validated for ELISA

Specifications

Antigen SFTS Virus HB29
Applications ELISA
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias B303_sMgp1, Membrane glycoprotein polyprotein
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid synthetic peptide near the center of human SFTS Virus HB29 Membrane Glycoprotein. The immunogen is located within amino acids 570 - 620 of SFTS Virus HB29. Amino Acid Squence: ITTGRALLPAVNPGQEA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 13231111
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.