Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SGC-CBP30

Catalog No. p-4816860
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent CBP/p300 BRD inhibitor

Potent CBP/p300 bromodomain (BRD) inhibitor (Kd values are 21 and 32 nM for CBP and p300 BRDs respectively). Exhibits 40-fold and 250-fold selectivity for CBP over the first BRD of BRD4 (BRD4(1)) and BRD4(2) respectively. Accelerates FRAP recovery in cells at a concentration of 1 μM.

Chemical Identifiers

CAS 1613695-14-9
Molecular Formula C28H33ClN4O3

Specifications

Quantity 10 mg
Formula Weight 509.04
Purity >98%
Molecular Weight (g/mol) 509.05
Chemical Name or Material SGC-CBP30
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.