Learn More
Description
A very polar nitroterephthalic acid modified PEG phase suited to the analysis of free fatty acids
- Nitroterephthalic acid modified PEG polar phase
- Ideal for low molecular weight acids
- Applications include volatile free acids, fatty acid methyl esters (FAMEs), alcohols, aldehydes and ketones
Specifications
Specifications
| For Use With (Application) | Ketones Analysis, Fatty Acid Methyl Esters, Alcohol Analysis, Aldehydes Analysis, Acrylates, Ketones |
| Film Thickness | 0.25 μm |
| Length (Metric) | 25 m |
| Packing Material | BP21 (FFAP) |
| Polarity | Polar |
| Coating | Fused Silica |
| USP Type | G25, G35 |
| Stationary Phase | Polyethylene Glycol |
| Diameter (Metric) Inner | 0.22 mm |
| Functionality/Interaction Type | Hydrogen Bonding or Dipole interactions |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
