Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trajan™ BP21 (FFAP) GC Capillary Columns

Click to view available options
Length (Metric):
25 m
30 m
50 m
60 m
Diameter (Metric) Inner:
0.22 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.25 μm
0.5 μm
1 μm
Description
A very polar nitroterephthalic acid modified PEG phase suited to the analysis of free fatty acids
- Nitroterephthalic acid modified PEG polar phase
- Ideal for low molecular weight acids
- Applications include volatile free acids, fatty acid methyl esters (FAMEs), alcohols, aldehydes and ketones
Specifications
Specifications
USP Type | G25, G35 |
Diameter (Metric) Inner | 0.25 mm |
Functionality/Interaction Type | Hydrogen Bonding or Dipole interactions |
Stationary Phase | Polyethylene Glycol |
Max. Temperature | 250°C |
For Use With (Application) | FAMEs Analysis, Fatty Acid Methyl Esters, Alcohol Analysis, Aldehydes Analysis, Acrylates, Ketones |
Temperature Range | 35°C to 240/250°C |
Film Thickness | 0.25 μm |
Length (Metric) | 60 m |
Packing Material | BP21 (FFAP) |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction