Learn More
MilliporeSigma™ Supelco™ SGE Gas Tight Syringes, Luer Lock
Catalog No. p-7213558 Shop All MilliporeSigma Supelco Products
Click to view available options
Volume (Metric):
50 μL
100 μL
250 μL
500 μL
1 mL
2.5 mL
5 mL
10 mL
25 mL
50 mL
100 mL
Can be effectively used for the introduction of samples to the gas sampling and injection system (GASIS) inlet
  • Maximum inertness for liquid or gas samples
  • Suitable for manual disperser/diluter applications
  • High accuracy of dispensed volume
  • Wide variety of terminations
  • Less time consuming
  • Improved work flow analysis
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Material Borosilicate Glass (Barrel), PTFE (Plunger)
Product Type SGE Gas Tight Syringe
Model 50 F-LL-GT
Quantity 1 Ea.
Volume (Metric) 50 μL
Connection Type Luer Lock, Kel-F Luer Cones
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.