Learn More
SGE Gastight™ Syringe, Fixed Needle (LTN), Sold by MilliporeSigma™ Supelco™
Catalog No. 111011052
Click to view available options
Volume (Metric):
10 μL
25 μL
50 μL
100 μL
250 μL
500 μL
1000 μL
Suitable for manual dispenser/diluter applications. Maximum inertness for liquid or gas samples.
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass (Barrel), PTFE (Plunger), Stainless Steel (Tip)
Needle Gauge 22 G
Diameter (Metric) Outer Needle 0.72 mm
Product Type SGE Gas Tight Syringe
Model 1 MDF-GT
Needle Point Number 2
Quantity 1 Ea.
Volume (Metric) 1000 μL
Length (Metric) Needle 50 mm
Needle Point Style Bevel
Connection Type Fixed
Show More Show Less
GASTIGHT is a registered trademark of Hamilton Co.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.