Learn More
MilliporeSigma™ Supelco™ SGE Needles for RN Syringes (Cone Tip)
Catalog No. p-7213559 Shop All MilliporeSigma Supelco Products
Click to view available options
Volume (Metric):
1 μL
For Use With (Equipment):
Removable Needle Syringe (1 B-7 C)
For easy replacement of needles that get damaged in the field. Needle is seated at zero graduation mark.
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel (Tip)
Needle Gauge 23 G
Diameter (Metric) Outer Needle 0.63 mm
Product Type SGE Needles
Model 1 B-7 C
Needle Point Number 1
Quantity 1 Ea.
Volume (Metric) 1 μL
Length (Metric) Needle 70 mm
Needle Point Style Cone
For Use With (Equipment) Removable Needle Syringe (1 B-7 C)
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.