Learn More
Trajan™ Replacement Plungers for eVol and eVol NMR Syringes
Click to view available options
For Use With (Equipment):
eVol NMR Syringe (5μL)
eVol NMR Syringe (50μL)
eVol NMR Syringe (500μL)
Description
- Replacement plungers for gas tight syringes
- Designed to match the specific syringe barrel to maintain a gas tight seal
Specifications
Specifications
Product Type | Replacement Plunger |
For Use With (Equipment) | eVol NMR Syringe (500μL) |