Learn More
MilliporeSigma™ Supelco™ SGE Syringe Valves Push button with Luer Lock
Catalog No. 111011379 Shop All MilliporeSigma Supelco Products
Click to view available options
For Use With:
Any Luer Lock Syringe
SGE 5-100 mL Syringe
Catalog No. 11-101-1379
Supplier: MilliporeSigma™ Supelco™ 509353
Only null left
Add to Cart
Add to Cart
Syringe valves are ideal for sample storage and transportation or to pressurize samples prior to injection.
SGE offers several types of manually-operated syringe valves: push-pull for smaller volumes (2.5 μL-2.5 mL) and push-button valves (50 μL-2 L). SGE Luer Tip, Luer Lock and removable needle syringes can be fitted with valves. The valve allows fluid to be stored in the syringe without contamination.Cat. No. 509337 is a push-pull valve with removable 0.63 mm O.D. x 5 cm cone tip needle. This valve will fit removable needle syringes, 25 μL-2.5 mL. Cat. No. 509345 is a push-pull with Kel-F male Luer lock termination to fit removable needle syringes, 25 μL-2.5 mLCat. No. 509353 is a push-button with Kel-F male Luer lock termination. This fits 5 mL to 2 liter syringes with removable Luer Lock or removable needle.Cat. No. 509361 is a Luer Lock valve for any Luer Lock and Luer Tip syringe (50 mul to 2 liter).
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 5-2,000 mL
Packaging Quantity 1/Pk.
For Use With SGE 5-100 mL Syringe
Material Stainless Steel
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.