Learn More
MilliporeSigma™ Supelco™ SGE Syringes, Removable Needle (RN)
Catalog No. p-7213542
Click to view available options
Volume (Metric):
0.5 μL
Needle Gauge:
23 G
For use with Agilent/HP 7673 and 7683 Autosamplers
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel (Tip)
Needle Gauge 23 G
Product Type Standard Syringe
Model 0.5 BR-HP-0.63
Needle Point Number 1
Quantity 1 Ea.
Volume (Metric) 0.5 μL
Length (Metric) Needle 42 mm
Needle Point Style Cone
For Use With (Equipment) Agilent/HP 7673 and 7683 Autosamplers
Connection Type Removable
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.