Learn More
SGE™ Terry Tool Tubing Cutter Replacement Part
SDP
Catalog No. SG082781 Shop All SGE Inc. Products
Change view
Click to view available options
Quantity:
3/Pk.
Catalog No. Quantity
SG-082781 3/Pk.
1 options
Catalog No. SG-082781 Supplier SGE™ Supplier No. 082781
Replacement cutting wheel for use with Terry-Tool Tubing Center.
  • Replacement wheel
  • For cutting 0.0625 in. and 0.125 in. stainless steel tubing
Specifications
Description Replacement cutting wheel
For Use With (Application) Terry-Tool Tubing Center
Quantity 3/Pk.