Learn More
SGE™ Tubing Cutter
SDP
Catalog No. SG082782 Shop All SGE Inc. Products
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
SG-082782 Stainless Steel
1 options
Catalog No. SG-082782 Supplier SGE™ Supplier No. 082782
For GC tubing
  • Made of stainless-steel
  • O.D.: 1/8 in.
Specifications
Product Type Tubing Cutter
For Use With (Equipment) GC tubing
Material Stainless Steel
For Use With (Application) Used to get right angle cuts with minimal burring or chipping of the tubing