Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SGEF Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB328005 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB328005 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB328005 Supplier Novus Biologicals Supplier No. NBP305912PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SGEF Polyclonal antibody specifically detects SGEF in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence

Specifications

Antigen SGEF
Applications Immunocytochemistry
Classification Polyclonal
Conjugate PE
Dilution Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias Rho guanine nucleotide exchange factor (GEF) 26
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SGEF fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 26084
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.