Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ SGI 1027
Click to view available options
Quantity:
10 mg
50 mg
Description
SGI 1027 is a DNA methyltransferase inhibitor. Inhibits the mammalian DNA methyltransferases DNMT3B, DNMT3A and DNMT1 (IC50 values are 7.5, 8 and 12.5 μM with Poly(dl-dC) as the substrate). Reactivates silenced tumor suppressor genes by reducing CpG island hypermethylation.
Specifications
Specifications
| Purity | >99% |
| Quantity | 10 mg |
| Product Type | SGI 1027 |
| Molecular Weight (g/mol) | 461.52 |
| Inhibitors | SGI 1027 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction