Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SGK1 Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB055426 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB055426 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB055426 Supplier Novus Biologicals Supplier No. NBP298778AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SGK1 Polyclonal specifically detects SGK1 in Human samples. It is validated for Western Blot.

Specifications

Antigen SGK1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.11, EC 2.7.11.1, serine/threonine protein kinase SGK, serine/threonine-protein kinase Sgk1, serum/glucocorticoid regulated kinase, serum/glucocorticoid regulated kinase 1, Serum/glucocorticoid-regulated kinase 1, SGK
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human SGK1.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 6446
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.