Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SGK1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP219010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP219010 0.1 mg
NBP219010SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP219010 Supplier Novus Biologicals Supplier No. NBP219010
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

SGK1 Polyclonal specifically detects SGK1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SGK1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml
Formulation PBS with 0.05% BSA with 0.05% Sodium Azide
Gene Alias EC 2.7.11, EC 2.7.11.1, serine/threonine protein kinase SGK, serine/threonine-protein kinase Sgk1, serum/glucocorticoid regulated kinase, serum/glucocorticoid regulated kinase 1, Serum/glucocorticoid-regulated kinase 1, SGK
Gene Symbols SGK1
Host Species Rabbit
Immunogen A portion of amino acids 360-410 from human SGK1 was used as the immunogen for this antibody. NP_005618.2.
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 6446
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, new world monkey, macaque, rhesus monkey, opossum, gibbon, orangutan, and chimpanzee, 95% in panda, Carolina anole, cow, dog, Chinese hamster, zebrafish, horse, mole rat, elephant, platypus, and pig, 90% in mouse, rat, zebra finch, frog, cichlid fish, rabbit, guinea pig, chicken, and wild turkey, 85% homologouus in salmon.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.