Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SGK1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$181.72 - $418.55
Specifications
| Antigen | SGK1 |
|---|---|
| Concentration | 0.5 mg/ml |
| Dilution | Western Blot 1-3 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP219010SS
![]() |
Novus Biologicals
NBP219010SS |
0.025 mg |
Each for $181.72
|
|
|||||
NBP219010
![]() |
Novus Biologicals
NBP219010 |
0.1 mg |
Each for $418.55
|
|
|||||
Description
SGK1 Polyclonal specifically detects SGK1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| SGK1 | |
| Western Blot 1-3 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.7.11, EC 2.7.11.1, serine/threonine protein kinase SGK, serine/threonine-protein kinase Sgk1, serum/glucocorticoid regulated kinase, serum/glucocorticoid regulated kinase 1, Serum/glucocorticoid-regulated kinase 1, SGK | |
| SGK1 | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen for this antibody is 100% homologous in human, new world monkey, macaque, rhesus monkey, opossum, gibbon, orangutan, and chimpanzee, 95% in panda, Carolina anole, cow, dog, Chinese hamster, zebrafish, horse, mole rat, elephant, platypus, and pig, 90% in mouse, rat, zebra finch, frog, cichlid fish, rabbit, guinea pig, chicken, and wild turkey, 85% homologouus in salmon. |
| 0.5 mg/ml | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, DNA Repair | |
| PBS with 0.05% BSA with 0.05% Sodium Azide | |
| 6446 | |
| A portion of amino acids 360-410 from human SGK1 was used as the immunogen for this antibody. NP_005618.2. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title