Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SGLT1/SLC5A1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220338 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220338 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220338 Supplier Novus Biologicals Supplier No. NBP220338
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SGLT1/SLC5A1 Polyclonal specifically detects SGLT1/SLC5A1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen SGLT1/SLC5A1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation PBS, 1% BSA, 20% Glycerol (pH7). with 0.025% Proclin 300
Gene Accession No. P13866
Gene Alias D22S675, High affinity sodium-glucose cotransporter, Na+/glucose cotransporter 1, NAGTsodium/glucose cotransporter 1, SGLT1Na(+)/glucose cotransporter 1, solute carrier family 5 (sodium/glucose cotransporter), member 1, Solute carrier family 5 member 1
Gene Symbols SLC5A1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SGLT1. The exact sequence is proprietary.
Molecular Weight of Antigen 73 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 6523
Test Specificity Species reactivity based on sequence identity: Pig (88%), Bovine (89%).
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.